di-Lithium tetraborate ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Dilithium tetraborate
, Lithium tetraborate
93-0355-500G
93-0355-100G
100200-724EA
101.91
USD
100200-724
100196-424
di-Lithium tetraborate ≥99%
di-Lithium tetraborate
Formula:
Li₂B₄O₇ MW: 169.12 g/mol Melting Pt: 917 °C Density: 1.4 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011083 CAS Number: 12007-60-2 EINECS: 234-514-3 Merck Index: 14,05524 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...