Trilithium orthophosphate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Lithium orthophosphate
, Lithium phosphate
, Lithium phosphate tribasic
93-0349-500G
93-0349-100G
100207-754EA
169.85
USD
100207-754
100208-484
Trilithium orthophosphate ≥98%
tri-Lithium orthophosphate
Formula:
Li₃O₄P MW: 115.79 g/mol Melting Pt: 1205 °C Density: 2,54 Storage Temperature: Ambient |
MDL Number:
MFCD00016187 CAS Number: 10377-52-3 EINECS: 233-823-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...