Lithium perchlorate trihydrate ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-0344-100G
93-0344-500G
100205-242EA
168.43
USD
100205-242
100205-240
Lithium perchlorate trihydrate ≥98%
Lithium perchlorate trihydrate
Formula:
ClLiO₄·3H₂O MW: 160.44 g/mol Melting Pt: 91.85 °C Density: 1.841 Storage Temperature: Ambient |
MDL Number:
MFCD00149765 CAS Number: 13453-78-6 EINECS: 232-237-2 UN: 1481 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...