Lithium molybdate(VI) ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Molybdic acid lithium saltdi-Lithium molybdateLithium molybdateLithium molybdenum oxide

93-0337-100G 93-0337-500G
100201-802EA 70.77 USD
100201-802 100210-674
Lithium molybdate(VI) ≥99%
Lithium molybdate(VI)
Formula: Li₂MoO₄
MW: 173.82 g/mol
Melting Pt: 705 °C
Density: 2,66
MDL Number: MFCD00016178
CAS Number: 13568-40-6
EINECS: 236-977-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR