Ammonium dihydrogen phosphate ≥98% ACS
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Ammonium dihydrogenphosphate
, Ammonium phosphate, monobasic
, Ammonium Phosphate Monobasic
93-0221-250G
93-0221-1000G
100206-866EA
171.26
USD
100206-866
100206-868
Ammonium dihydrogen phosphate ≥98% ACS
Ammonium dihydrogen phosphate
Formula:
NH₄H₂PO₄ MW: 115.03 g/mol Melting Pt: 190 °C Density: 1.803 Storage Temperature: Ambient |
MDL Number:
MFCD00003396 CAS Number: 7722-76-1 EINECS: 231-764-5 Merck Index: 13,00547 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...