Dithallium trioxide ≥99.99% (Tl basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Danger
|
81-8100-50G
81-8100-10G
100206-136EA
450.09
USD
100206-136
100206-858
Dithallium trioxide ≥99.99% (Tl basis), PURATREM
Dithallium trioxide
Formula:
O₃Tl₂ MW: 456.76 g/mol Melting Pt: ∼717 °C Density: 5.56 g/cm³ (0 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011276 CAS Number: 1314-32-5 EINECS: 215-229-3 UN: 1707 ADR: 6.1,II Merck Index: 12,09404 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...