Iridium ≥99.7%, wire
Supplier: STREM CHEMICALS INC MS
77-2700-50CM
77-2700-10CM
100210-088EA
343.94
USD
100210-088
100198-256
Iridium ≥99.7%, wire
Iridium
Formula:
Ir MW: 192.22 g/mol Boiling Pt: 4130 °C Melting Pt: 2410 °C Density: 22,421 |
MDL Number:
MFCD00011062 CAS Number: 7439-88-5 EINECS: 231-095-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...