Combination Carbon Monoxide and Smoke Alarm, KIDDE, ORS Nasco
Supplier: ORS Nasco
5011133143
300055-002EA 85.2 USD
300055-002
Combination Carbon Monoxide and Smoke Alarm, KIDDE, ORS Nasco
Smoke Detectors
This alarm has two LEDs, test/reset button and battery safeguard.
  • Talking alarm announces hazard type detected
  • Temperature range min. 40 °F, max. 100 °F
  • 85 dB sound level
  • Low-battery warning
  • For use with 3 AA batteries
Weight: 4.31 lbs.
Ordering information: Batteries to be ordered separately.