Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Protease Assay Kit, G-Biosciences

Supplier: G-Biosciences
786-028
The Protease Assay Kit is designed for the quantitative determination of proteases present in protein sample, using a dye-labeled protein substrate

Chemically stabilized Trypsin (MSG-Trypsin™) is supplied with the kit as a general protease standard; however, other specific protease standards can also be used. MSG-Trypsin™ is an ultra-pure trypsin from bovine pancreas, modified by methylation followed by TPCK treatment and is extremely resistant to autolysis. The kit components are sufficient for 50 assays in a microtiter plate format or 0.5ml assay tubes.

Accessories: Resorufin-Casein Protease Substrate (#786-321): A colorimetric substrate that when treated with proteases releases resorufin that has an absorbance of 570nm. Supplied lyophilized. FITC-Casein Protease Substrate (#786-322): A fluorescent (fluorescein isothiocyanate (FITC)) substrate that when treated with proteases releases FITC that has an excitation at 485nm and emission at 535nm. Supplied lyophilized.

The proteases present in the sample of interest will digest the protein substrate and release dye labeled peptides. The absorbance of the dye-labeled peptide is measured at 570nm for determination of protease activity.

Ordering information: Accessories not included.




 

ORDER

Expand All / Collapse All
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
Protease Assay Kit 50 assays 786-028 82023-260 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
FITC-Casein Protease Substrate 1 mg 786-322 78000-046 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Resorufin-Casein Protease Substrate 4 mg 786-321 78000-044 Each (4mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty