Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PURELAB® Ultra Water Purification Systems, ELGA LabWater

Supplier: ELGA
PURELAB®
ULXXXANM2 ULXXXSCM2 ULXXXGEM2/V LC109 ULXXXIOM2/V ULXXXBSM2 LC118 LC105 ULXXXANM2/V LC134 LC197 ULXXXIOM2 ULXXXSCM2/V LC184 LC151 LC185 ULXXXBSM2/V LC182 LC183 ULXXXGEM2
The PURELAB Ultra is the intelligent Type I+ ultrapure water purification system for your most critical applications

  • Twin purification pack “PureSure” system gurantees water purity
  • Stops organics leaching in to the final water purity
  • Real-time TOC monitoring ensures confidence in organic quality (Genetic and Analytic models only)
  • PureSure system ensures that work is not interrupted by cartridge replacement
  • Ongoing bacterial performance guaranteed through complete sanitization of all wetted parts

Suggested applications include mass spectrometry, molecular biology, ultra trace analyses, electrochemistry, atomic spectroscopy, liquid chromatography, ion chromatography, cell cultures, qualitative analyses, gas chromatography, immunochemistry.

It incorporates many unique and innovative features that set it apart from other systems and guarantees high water purity of 18.2 MΩ-cm.

Certifications: UL and C-UL listed. CSA certified. CE marked.

Accessories information: Additional accessories are available; please contact your VWR representative for additional information.

Ordering information: Feed water must be from a pre-treated source such as deionized, distilled or reverse osmosis. Two of the same purification packs required and sold separately and if required, a point of use filter. Validation labor not included with systems that include validation documentation. If the feed pressure exceeds 10 psi, a pressure regulator is required.




 

ORDER

Expand All / Collapse All
Description Water Quality Dispense Rate Supplier No. VWR Catalog Number Unit Price Quantity
PURELAB® Ultra Analytic Ultra-Pure Polishing System Type I+ 2.0 L/min max ULXXXANM2 89204-052 Each Retrieving
Product available on GSA Advantage®
PURELAB® Ultra Analytic Ultra-Pure Polishing System with Validation Documentation Type I+ 2.0 L/min max ULXXXANM2/V 89204-054 Each Retrieving
Product available on GSA Advantage®
PURELAB® Ultra Bioscience Ultra-Pure Polishing System Type I+ 2.0 L/min max ULXXXBSM2 89204-056 Each Retrieving
Product available on GSA Advantage®
PURELAB® Ultra Bioscience Ultra-Pure Polishing System with Validation Documentation Type I+ 2.0 L/min max ULXXXBSM2/V 89204-058 Each Retrieving
Product available on GSA Advantage®
PURELAB® Ultra Genetic Ultra-Pure Polishing System Type I+ 2.0 L/min max ULXXXGEM2 89204-060 Each Retrieving
Product available on GSA Advantage®
PURELAB® Ultra Genetic Ultra-Pure Polishing System with Validation Documentation Type I+ 2.0 L/min max ULXXXGEM2/V 89204-062 Each Retrieving
Product available on GSA Advantage®
PURELAB® Ultra Ionic Ultra-Pure Polishing System Type I+ 2.0 L/min max ULXXXIOM2 89204-064 Each Retrieving
Product available on GSA Advantage®
PURELAB® Ultra Ionic Ultra-Pure Polishing System with Validation Documentation Type I+ 2.0 L/min max ULXXXIOM2/V 89204-066 Each Retrieving
Product available on GSA Advantage®
PURELAB® Ultra Scientific Ultra-Pure Polishing System Type I+ 2.0 L/min max ULXXXSCM2 89204-068 Each Retrieving
Product available on GSA Advantage®
PURELAB® Ultra Scientific Ultra-Pure Polishing System with Validation Documentation Type I+ 2.0 L/min max ULXXXSCM2/V 89204-070 Each Retrieving
Product available on GSA Advantage®

PURELAB® Ultra Consumables
Description Supplier No. VWR Catalog Number Unit Price Quantity
Point-of-Use Biofilter LC197 89204-398 Each Retrieving
Product available on GSA Advantage®
ENDOGARD 5000 MWCO Cartridge LC151 89204-344 Each Retrieving
Product available on GSA Advantage®
LABPURE S2 Purification Cartridge, SDI Feed LC183 89204-370 Each Retrieving
Product available on GSA Advantage®
LABPURE S3 Purification Cartridge, Low Ionic Feed LC184 89204-372 Each Retrieving
Product available on GSA Advantage®
LABPURE S4 Purification Cartridge, Low TOC Feed LC185 89204-374 Each Retrieving
Product available on GSA Advantage®
Ultra Microfilter, 0.05 µm LC109 89204-290 Each Retrieving
Product available on GSA Advantage®
Point-of-Use Microfilter, 0.2 µm LC134 89204-328 Each Retrieving
RO S1 Purification Cartridge LC182 89204-368 Each Retrieving
UV Lamp, 185 nm LC118 89204-326 Each Retrieving
Product available on GSA Advantage®
UV Lamp, 254 nm LC105 89204-286 Each Retrieving
Product available on GSA Advantage®
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Model PURELAB® Ultra Genetic PURELAB® Ultra Scientific PURELAB® Ultra Ionic PURELAB® Ultra Bioscience PURELAB® Ultra Scientific PURELAB® Ultra Analytic PURELAB® Ultra Genetic PURELAB® Ultra Ionic PURELAB® Ultra Analytic PURELAB® Ultra Bioscience
Toc Content < 1 ppb 3-10 ppb < 1 ppb 3-10 ppb < 1 ppb 3-10 ppb
Bacteria <0.1 CFU/mL with POU filter fitted
Particles Ultrafiltration Optional 0.2 µm POU filter 0.05 µm Ultrafiltration Optional 0.2 µm POU filter 0.05 µm Ultrafiltration 0.05 µm Ultrafiltration
Inorganics @ 25°C 18.2 MΩ.cm
Endotoxin <0.001 EU/mL <0.001 EU/mL <0.001 EU/mL <0.001 EU/mL

ACCESSORIES

Expand All / Collapse All

Accessories
Description Supplier No. VWR Catalog Number Unit Price Quantity
Inlet Pressure Regulator LA652 89204-220 Each Retrieving
Closed Circuit Ringmain Kit LA647 89204-216 Each Retrieving
Open Circuit Ringmain Kit LA646 89204-214 Each Retrieving
Remote Control Station LA645 89204-212 Each Retrieving
Integral Dispense Gun LA644 89204-210 Each Retrieving
Remote Dispense Station and Dispense Gun LA643 89204-208 Each Retrieving
Ultra/Classic Cartridge for PURELAB® Chorus LA698-US 75803-128 Each Retrieving
Wall Mounting Kit LA622 89204-198 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SERVICES


Services: ELGA LabWater provides several service options such as Extended Warranty, Installation, Preventative Maintenance, and Validation. To learn more about these services please search "Elga Water Technical Service" in the VWR search field.

In addition, VWRCATALYST™ can provide an array of services such as on-site IQ, OQ, PQ, Customized Protocol Generation, Equipment Calibration, Asset Management, and Certification Services (US only). Please call 888-793-2300 or email VWRCATALYST™@vwr.com for further details.

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty