PURELAB® Option R7 and R15 Water Purification Systems, ELGA LabWater
OR007BPM1/V OR015BPM1/V OR015BPM1/V OR007BPM1 OR015BPM1 OR007BPM1 OR015BPM1 OR007BPM1/V
89204-000EA 7910.59 USD
CA89204-002 CA89204-010 89204-000 CA89204-000 89204-002 89204-010 89204-008 CA89204-008
PURELAB® Option R7 and R15 Water Purification Systems, ELGA LabWater
The PURELAB Option provides Type II+ pure water on demand with a wide range of flow-rates to suit your needs
  • Delivers market leading microbial performance
  • Features ‘Reverse Osmosis feed optimized’ resin mixes
  • Increases the ion exchange capacity of consumables and minimize running costs
  • Quick and easy sanitization and replacement of consumables to reduce maintenance time
In addition to simple sanitization and composite vent filtration, the PURELAB Option-R system is the only fully recirculating Type II+ pure water system on the market, ensuring high specification water. PURELAB Option systems are designed to be easy to access whether wall or bench mounted with a convenient dispense tap. They can be used with our wrap-around reservoirs to save space whilst optimizing purity.
Applications range from glassware washing and the feeding of ultrapure water systems to cell culture and media preparation. The PURELAB Option-R provides additional bacteria and inorganic quality for your more critical applications.
Certifications: UL and C-UL listed. CSA certified. CE marked.
Accessories information: Additional accessories are available; contact your VWR representative for more information.
Ordering information: If feedwater pressure is above 2 bar (30 psi) a pressure regulator will be required. A storage reservoir is required for this system to operate; these are sold separately. Unit supplied with mandatory consumables for standard operation. If required, optional point of use filters can be purchased. Validation labor not included with systems that include validation documentation.