Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PURELAB® Option S7 and S15 Water Purification Systems, ELGA LabWater

Supplier: ELGA
PURELAB®
LA611 LA613 OS015BPM1/V LA612 OS015BPM1 LA620 LA592 LC123 LA591 LC143 LC140 LC141 LC136M2 OS007BPM1 OS007BPM1/V
The PURELAB Option provides Type II pure water on demand with a wide range of flow-rates to suit your needs

  • Features ‘Reverse Osmosis feed optimized’ resin mixes
  • Increase the ion exchange capacity of consumables and minimize running costs
  • Quick and replacement of consumables to reduce maintenance time

PURELAB Option systems are designed to be easy to access whether wall or bench mounted with a convenient dispense tap. They can be used with our wrap-around reservoirs to save space whilst optimizing purity.

Applications range from glassware washing and the feeding of ultrapure water systems to cell culture and media preparation

Certifications: UL and C-UL listed. CSA certified. CE marked.

Accessories information: Additional accessories are available; contact your VWR representative for more information.

Ordering information: If feedwater pressure is above 2 bar (30 psi) a pressure regulator will be required. A storage reservoir is required for this system to operate; these are sold separately. Unit supplied with mandatory consumables for standard operation. Validation labor not included with systems that include validation documentation.




 

ORDER

Expand All / Collapse All
Description Water Quality Dispense Rate Daily Output Make Up Flow Rate Supplier No. VWR Catalog Number Unit Price Quantity
PURELAB® Option S 7 Water Purification System with Raw Water Boost Pump Type II From Reservoir 180 L/24hr 7.5 LPH (180 L/day) OS007BPM1 89203-984 Each Retrieving
Product available on GSA Advantage® UL Listed cUL Listed CSA Certified
PURELAB® Option S 7 Water Purification System with Raw Water Boost Pump and Validation Documentation Type II From Reservoir 180 L/24hr 7.5 LPH (180 L/day) OS007BPM1/V 89203-986 Each Retrieving
Product available on GSA Advantage® UL Listed cUL Listed CSA Certified
PURELAB® Option S 15 Water Purification System with Raw Water Boost Pump Type II From Reservoir 360 L/24hr 15 LPH (360 L/day) OS015BPM1 89203-992 Each Retrieving
Product available on GSA Advantage® UL Listed cUL Listed CSA Certified
PURELAB® Option S 15 Water Purification System with Raw Water Boost Pump and Validation Documentation Type II From Reservoir 360 L/24hr 15 LPH (360 L/day) OS015BPM1/V 89203-994 Each Retrieving
Product available on GSA Advantage® UL Listed cUL Listed CSA Certified

PURELAB® Option S Consumables
Description Supplier No. VWR Catalog Number Unit Price Quantity
RO Membranes LC143 89204-284 Each Retrieving
Product available on GSA Advantage®
Twin Pack Cartridge LC141 89204-308 Each Retrieving
Product available on GSA Advantage®
Pre-Treatment Cartridge, Removes Particulates and Chlorine LC140 89204-302 Each Retrieving
Pre-Treatment Cartridge, Removes Particulates and Free Chlorine LC123 89204-310 Each Retrieving
Product available on GSA Advantage®

PURELAB® Water Storage Reservoirs
Description Supplier No. VWR Catalog Number Unit Price Quantity
Docking Tank, DV35 LA620 89204-194 Each Retrieving
Product available on GSA Advantage®
Hydrophobic Microporous Composite Vent Filter LC136M2 89204-342 Each Retrieving
Stand-Alone Storage Reservoir, 25 l LA611 89204-178 Each Retrieving
Product available on GSA Advantage®
Stand-Alone Storage Reservoir, 40 l LA612 89204-182 Each Retrieving
Product available on GSA Advantage®
Stand-Alone Storage Reservoir, 75 l LA613 89204-186 Each Retrieving
Product available on GSA Advantage®
Wall Mounting Kit for 25/35/40 l Reservoir LA591 89204-200 Each Retrieving
Product available on GSA Advantage®
Wall Mounting Kit for 75 l Reservoir LA592 89204-202 Each Retrieving
Product available on GSA Advantage®
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Inorganics 1 to >10 MΩ.cm
Toc Content <30 ppb

ACCESSORIES

Expand All / Collapse All

Accessories
Description Supplier No. VWR Catalog Number Unit Price Quantity
Feedwater Pressure Regulator LA512 89204-218 Each Retrieving
Pre-Filter Assembly Kit (if boost pump fitted) LA582 89508-460 Each Retrieving
Installation Kit for Small Box Products, Multi-Purpose LA513 89508-456 Each Retrieving
Filter Housing, 10" LA518 89204-450 Each Retrieving
OPTION S Flow Upgrade Kit (7–15 L/hr) LA604 89508-454 Each Retrieving
RO Flow Kit Upgrade LA605 89204-222 Each Retrieving
Wall Mounting Kit LA622 89204-198 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SERVICES


Services: ELGA LabWater provides several service options such as Extended Warranty, Installation, Preventative Maintenance, and Validation. To learn more about these services please search "Elga Water Technical Service" in the VWR search field.

In addition, VWRCATALYST™ can provide an array of services such as on-site IQ, OQ, PQ, Customized Protocol Generation, Equipment Calibration, Asset Management, and Certification Services (US only). Please call 888-793-2300 or email VWRCATALYST™@vwr.com for further details.

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty