Side Load Glove Box Holder, Mitchell Plastics
MGPC-1000B MGPCLL-2000W MGPC-3000B MGPC-3000W MGPCLL-3000B MGPC-2000W MGPCLL-2000B
89140-652EA 208.77 USD
89140-652 89140-664 89140-650 89140-656 89140-668 89140-670 89140-660
Side Load Glove Box Holder, Mitchell Plastics
Most all glove boxes may be used with this holder
  • Load from the side
  • Mount vertically or horizontally
  • Two holes and double sided tape provided for mounting
They provide extra support for removal of lightweight gloves from their cardboard boxes. The holders may be mounted in the most convenient locations for easy access.
Ordering information: There is a minimum order requirement of 25 for this item.