Would you like to log in again?
Sentino® Microbiology Pump, Pall Laboratory
Supplier: Pall Laboratory
Sentino®
13180 13186 13186 13180
CA89183-082 10841-650 CA10841-650 89183-082
Sentino® Microbiology Pump, Pall Laboratory
The Sentino® Microbiology Pump is intended for microbiological evaluations of aqueous samples by Membrane Filter (MF) Technique.
  • Peristaltic action
  • Compact pump
  • Minimizes maintenance
This unique pump is designed to use peristatlic action to draw the sample through the filter and send filtrate directly to drain or waste.
The disposable Fluid Path is made specifically for use with the Sentino® Microbiology Pump. The unique peristaltic action of the Sentino® Pump is designed around a disposable Fluid Path that can easily be replaced before biofilm build-up can cause contamination.
The compact Sentino® Microbiology Pump simplifies the time it takes to prepare for membrane filtrations, frees up more lab space, simplifies testing, minimizes equipement maintenance, and provides a level of contamination control often overlooked within a lab.
Certifications: CE marked.
Ordering information: 100–240 VAC, 47–63 Hz.
Order Now
ORDER
ACCESSORIES
COMPLEMENTARY PRODUCTS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty