Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Exicycler Real-Time PCR System, Bioneer

Supplier: BIONEER, INC.
Exicycler 96
A-2060
Exicycler is a flexible, 5-color system with advanced features such as a thermal gradient function as well as light tunnel technology which eliminates the need for a reference dye.

Bioneer's Exicycler 96 is different from most Real Time systems in that we've addressed a significant problem that affects many systems – edge effects and the need for reference dyes. Most systems illuminate the wells from above which results in up to a 30% difference in light intensity from the center of the plate to the edges. Bioneer's Exicycler 96 uses our exclusive light tunnel technology to deliver the same number of photons to each well, eliminating the need to use reference dyes to adjust for intensity differences!

Exicycler 96 also offers several other features such as a thermal gradient for optimization. This is a critical feature to have for optimization of real time PCR and absolutely essential for multiplex optimization. Exicycler 96 has all the analysis modules you would expect in a Real-Time system including; Absolute Quantification, Relative Quantification, +/- Assays, SNP Genotyping, Melt Curve Analysis, etc. By using a high-sensitivity 2D 16-bit CCD sensor and high-quality optics, Exicycler 96 is able to acquire clean and sensitive signals from all the wells simultaneously. All fluorescent data is saved regardless of how the experiment was set up, which means that if a well was accidentally left out, the data can be recovered, or rather, rediscovered!




 

ORDER


The main product you are looking for is no longer available for purchase. Accompanying products and services may be found elsewhere on the page.

SPECIFICATIONS

Temperature Uniformity
Heating Rate
Gradient
No. of Channels

ACCESSORIES

Expand All / Collapse All

Accessories
Description Units Supplier No. VWR Catalog Number Unit Price Quantity
8-STRIP PCR TUBES WHT 0.2 mL Pack of 250 3111-50 97060-790 Pack of 250 Retrieving
PCR PLATE 96W SEMI-SKIRT WHT Pack of 25 3111-52 97060-792 Pack of 25 Retrieving
QPCR ADHESIVE SEALING FILM Each 3111-41 71002-570 Each Retrieving Restricted
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty