1,1,1-Trifluoroiodoethane ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 1-Iodo-2,2,2-trifluoroethane1,1,1-Trifluoro-2-iodoethane2,2,2-Trifluoroiodoethane2,2,2-Trifluoroethyliodide2-Iodo-1,1,1-trifluoroethane

09-7370-5G 09-7370-25G
100207-550EA 42.46 USD
100207-550 100199-750
1,1,1-Trifluoroiodoethane ≥99%
1,1,1-Trifluoroiodoethane

Organo Fluorine

Formula: C₂H₂F₃I
MW: 209.94 g/mol
Boiling Pt: 55…56 °C
Density: 2,130
MDL Number: MFCD00001082
CAS Number: 353-83-3
EINECS: 206-541-0
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR