Perfluoroheptanoic acid ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Perfluoroenanthic acidTridecafluoroheptylic acidTridecafluoroenanthic acidTridecafluoroheptanoic acid

09-6034-25G 09-6034-5G
100209-620EA 460 USD
100209-620 100209-622
Perfluoroheptanoic acid ≥97%
Perfluoroheptanoic acid

Organo Fluorine

Formula: C₇HF₁₃O₂
Boiling Pt: 78…80 °C (14 mmHg)
MDL Number: MFCD00039604
CAS Number: 375-85-9
EINECS: 206-798-9
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR