Pentafluorobenzaldehyde ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 2,3,4,5,6-Pentafluorobenzaldehyde

09-5440-1G 09-5440-25G 09-5440-5G
100197-350EA 45.29 USD
100197-350 100203-928 100201-412
Pentafluorobenzaldehyde ≥98%
Pentafluorobenzaldehyde

Organo Fluorine

Formula: C₇HF₅O
Boiling Pt: 164…166 °C
Melting Pt: 20 °C
Density: 1,588
Storage Temperature: Ambient
MDL Number: MFCD00003303
CAS Number: 653-37-2
EINECS: 211-502-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR