Pentafluoroiodobenzene ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Iodopentafluorobenzene

09-4950-5G 09-4950-25G
100205-820EA 232.12 USD
100205-820 100209-608
Pentafluoroiodobenzene ≥97%
Pentafluoroiodobenzene

Organo Fluorine

Formula: C₆F₅I
MW: 293.96 g/mol
Boiling Pt: 161 °C
Melting Pt: –29 °C
Density: 2,204
MDL Number: MFCD00001032
CAS Number: 827-15-6
EINECS: 212-565-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR