1-Fluoro-2-nitrobenzene

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: o-Fluoronitrobenzene2-Fluoronitrobenzene

09-3890-50G 09-3890-10G
100203-920EA 50.95 USD
100203-920 100207-958
1-Fluoro-2-nitrobenzene
1-Fluoro-2-nitrobenzene

Organo Fluorine

Formula: C₆H₄FNO₂
MW: 141.1 g/mol
Boiling Pt: 214…216 °C
Melting Pt: –8 °C
Density: 1,338
Flash Pt: 202 °F
Storage Temperature: Ambient
MDL Number: MFCD00007048
CAS Number: 1493-27-2
EINECS: 216-088-0
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR