Ethyl trifluoroacetate ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Trifluoroacetic ethyl ester
09-3270-500G
09-3270-100G
09-3270-25G
100204-842EA
77.85
USD
100204-842
100202-740
100204-844
Ethyl trifluoroacetate ≥99%
Ethyl trifluoroacetate
Organo Fluorine
Formula:
C₄H₅F₃O₂ MW: 142.08 g/mol Boiling Pt: 62 °C Melting Pt: –78 °C Density: 1,194 Flash Pt: 30 °F Storage Temperature: Refrigerator |
MDL Number:
MFCD00000419 CAS Number: 383-63-1 EINECS: 206-851-6 UN: 2924 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...