1,1-Dichloro-2,2-difluoroethylene ≥97%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
1,1-Dichlorodifluoroethylene
09-2360-100G
09-2360-25G
100205-818EA
2140.06
USD
100205-818
100202-892
1,1-Dichloro-2,2-difluoroethylene ≥97%
1,1-Dichloro-2,2-difluoroethylene
Organo Fluorine
Formula:
C₂Cl₂F₂ MW: 132.92 g/mol Boiling Pt: 19 °C Melting Pt: –116 °C Density: 1,439 |
MDL Number:
MFCD00039338 CAS Number: 79-35-6 EINECS: 201-198-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...