Chloropentafluorobenzene ≥99%

Supplier: STREM CHEMICALS INC MS

Synonyms: Pentafluorochlorobenzene

09-2010-25G 09-2010-100G
100204-488EA 124.55 USD
100204-488 100208-770
Chloropentafluorobenzene ≥99%
Chloropentafluorobenzene

Organo Fluorine

Formula: C₆ClF₅
MW: 202.51 g/mol
Boiling Pt: 122…123 °C (750 mmHg)
Melting Pt: –15 °C
Density: 1,568
Storage Temperature: Ambient
MDL Number: MFCD00000534
CAS Number: 344-07-0
EINECS: 206-450-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR