Chloropentacarbonylrhenium ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Pentacarbonylchlororhenium(I)Chloropentacarbonylrhenium(I)Rhenium pentacarbonyl chloride

75-4000-5G 75-4000-1G
100196-356EA 634.09 USD
100196-356 100196-354
Chloropentacarbonylrhenium ≥98%
Chloropentacarbonylrhenium
Formula: C₅ClO₅Re
MW: 361.71 g/mol
MDL Number: MFCD00013296
CAS Number: 14099-01-5
EINECS: 237-948-1
UN: 3466
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR