Potassium perrhenate ≥99% (Re basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
75-0500-10G
75-0500-2G
100198-252EA
578.89
USD
100198-252
100210-068
Potassium perrhenate ≥99% (Re basis)
Potassium perrhenate
Formula:
KO₄Re MW: 289.3 g/mol Boiling Pt: ~1365 °C Melting Pt: 555 °C Density: 4,887 |
MDL Number:
MFCD00011365 CAS Number: 10466-65-6 EINECS: 233-953-8 UN: 1479 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...