Hafnium ≥99.97% (Zr-3.1%), wire

72-0085-100CM 72-0085-25CM
100202-418EA 173.25 USD
100202-418 100204-098
Hafnium ≥99.97% (Zr-3.1%), wire
Hafnium
Formula: Hf
MW: 178.49 g/mol
Boiling Pt: 4602 °C
Melting Pt: 2227 °C
Density: 13,31
MDL Number: MFCD00011032
CAS Number: 7440-58-6
EINECS: 231-166-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR