1-Hexyl-3-methylimidazolium tetrafluoroborate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: [HMIM] [BF4]1-n-Hexyl-3-methylimidazolium tetrafluoroborate

07-0968-5G
100202-610EA 215.14 USD
100202-610
1-Hexyl-3-methylimidazolium tetrafluoroborate ≥98%
1-Hexyl-3-methylimidazolium tetrafluoroborate

Ionic Liquids

Formula: C₁₀H₁₉BF₄N₂
MW: 254.08 g/mol
Boiling Pt: ∼200 °C (1013 hPa)
Melting Pt: –81 °C
Density: 1,149
Flash Pt: >100 °C
MDL Number: MFCD03095459
CAS Number: 244193-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR