Ytterbium(III) chloride hydrate ≥99.99% (Yb basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

70-7003-25G 70-7003-5G
100203-258EA 345.35 USD
100203-258 100205-190
Ytterbium(III) chloride hydrate ≥99.99% (Yb basis), PURATREM
Ytterbium (III) chloride hydrate
Formula: Cl₃Yb·H₂O
MW: 297.41 g/mol
Density: 2.575
MDL Number: MFCD00149885
CAS Number: 19423-87-1
EINECS: 233-800-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR