1-Butyl-2,3-dimethylimidazoliumtetrafluoroborate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: [BDiMIM] [BF4]1-Butyl-2,3-dimethylimidazolium Tetrafluoroborate

07-0075-5G
100200-234EA 178.34 USD
100200-234
1-Butyl-2,3-dimethylimidazoliumtetrafluoroborate ≥98%
1-Butyl-2,3-dimethylimidazolium tetrafluoroborate

Ionic Liquids

Formula: C₉H₁₇BF₄N₂
MW: 240.05 g/mol
Density: 1,198
MDL Number: MFCD03427618
CAS Number: 402846-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR