Cerium ≥99.9%, chips
Supplier: STREM CHEMICALS INC MS
|
Danger
|
58-0040-100G
58-0040-25G
100201-008EA
1115.32
USD
100201-008
100204-636
Cerium ≥99.9%, chips
Cerium
Formula:
Ce MW: 140.12 g/mol Boiling Pt: 3257 °C Melting Pt: 815 °C Density: 6,90 |
MDL Number:
MFCD00010924 CAS Number: 7440-45-1 EINECS: 231-154-9 UN: 1333 ADR: 4.3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...