Barium manganate ≥90%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Barium manganate(VI)
56-7075-50G
56-7075-10G
100201-706EA
121.72
USD
100201-706
100202-770
Barium manganate ≥90%
Barium manganate
Formula:
BaMnO₄ Density: 4.850 |
MDL Number:
MFCD00011514 CAS Number: 7787-35-1 EINECS: 232-109-6 UN: 1479 ADR: 5.1,II Merck Index: 13,00984 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...