Tellurium tetrabromide ≥99.9% (Te basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Tellurium(IV) bromide
52-7000-50G
52-7000-10G
100202-674EA
726.09
USD
100202-674
100198-220
Tellurium tetrabromide ≥99.9% (Te basis)
Tellurium tetrabromide
Formula:
Br₄Te MW: 447.22 g/mol Melting Pt: 363 °C Density: 4.3 g/cm³ (20 °C) |
MDL Number:
MFCD00049575 CAS Number: 10031-27-3 EINECS: 233-090-7 UN: 1759 ADR: 8,II Merck Index: 12,09278 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...