Bis(tributyltin) oxide ≥96%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Bis(tri-n-butyltin)oxide
, TBTO
50-0700-100G
50-0700-500G
100196-302EA
59.45
USD
100196-302
100198-214
Bis(tributyltin) oxide ≥96%
Bis(tributyltin) oxide
Formula:
C₂₄H₅₄OSn₂ MW: 596.11 g/mol Boiling Pt: 254 °C (50 mmHg) Density: 1,17 (25°) Flash Pt: >230 °F Storage Temperature: Ambient |
MDL Number:
MFCD00009418 CAS Number: 56-35-9 EINECS: 200-268-0 UN: 2788 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...