Rhodium(III) chloride, anhydrous
Supplier: STREM CHEMICALS INC MS
45-1878-5G
45-1878-250MG
45-1878-1G
100206-692EA
762.89
USD
100206-692
100204-064
100204-062
Rhodium(III) chloride, anhydrous
Rhodium (III) chloride
Formula:
Cl₃Rh MW: 209.26 g/mol Melting Pt: 450 °C (dec.) Density: 5.38 g/cm³ (20 °C) |
MDL Number:
MFCD00011204 CAS Number: 10049-07-7 EINECS: 233-165-4 Merck Index: 12,08355 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...