Niobium ≥99.95% (metals basis excluding Ta), wire
Supplier: STREM CHEMICALS INC MS
|
Warning
|
41-0080-25CM
41-0080-100CM
100203-128EA
73.6
USD
100203-128
100208-426
Niobium ≥99.95% (metals basis excluding Ta), wire
Niobium
Formula:
Nb MW: 92.91 g/mol Boiling Pt: 4742 °C Melting Pt: 2468 °C Density: 8,57 |
MDL Number:
MFCD00011126 CAS Number: 7440-03-1 EINECS: 231-113-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...