Niobium ≥99.8%, foil

Supplier: STREM CHEMICALS INC MS
Warning

41-0055-100X50MM 41-0055-100X200MM
100205-112EA 77.85 USD
100205-112 100196-210
Niobium ≥99.8%, foil
Niobium
Formula: Nb
MW: 92.91 g/mol
Boiling Pt: 4742 °C
Melting Pt: 2468 °C
Density: 8,57
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR