Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triphenylarsine ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

33-4000.5 33-4000.25
100208-414 100208-412
Triphenylarsine ≥97%


Formula: C₁₈H₁₅As
MW: 306,24 g/mol
Boiling Pt: Min. 360 °C (1013 hPa)
Melting Pt: 58 °C
Density: 1.306 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00002994
CAS Number: 603-32-7
EINECS: 210-032-9
UN: 1557
ADR: 6.1,III
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty