Germanium(IV) chloride ≥99.9999% (Ge basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Germanium tetrachloride
, Tetrachlorogermane
32-0700-100G
32-0700-500G
32-0700-25G
100200-674EA
1050.22
USD
100200-674
100206-578
100207-656
Germanium(IV) chloride ≥99.9999% (Ge basis), PURATREM
Germanium (IV) chloride
Formula:
Cl₄Ge MW: 214.42 g/mol Boiling Pt: 83.1 °C Melting Pt: –49.5 °C Density: 1.880 Storage Temperature: Ambient |
MDL Number:
MFCD00011029 CAS Number: 10038-98-9 EINECS: 233-116-7 UN: 1760 ADR: 8,II Merck Index: 12,04418 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...