Zinc bromide ≥99.9% (Zn basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Zinc (II) bromide
30-3032-250G
30-3032-50G
100205-094EA
76.43
USD
100205-094
100208-880
Zinc bromide ≥99.9% (Zn basis)
Zinc bromide
Formula:
Br₂Zn MW: 225.2 g/mol Boiling Pt: 650 °C Melting Pt: 394 °C Density: 4.201 Storage Temperature: Ambient |
MDL Number:
MFCD00011294 CAS Number: 7699-45-8 EINECS: 231-718-4 UN: 1759 ADR: 8,II Merck Index: 12,10258 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...