Copper ≥99%, turnings

Supplier: STREM CHEMICALS INC MS

29-0090-500G 29-0090-100G
100197-480EA 128.8 USD
100197-480 100197-478
Copper ≥99%, turnings
Copper
Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8.92
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR