Nickel ≥99%, foil
Supplier: STREM CHEMICALS INC MS
|
Warning
|
28-0002-500CM
28-0002-20CM
28-0002-100CM
100196-936EA
1732.43
USD
100196-936
100196-934
100206-552
Nickel ≥99%, foil
Nickel
Formula:
Ni MW: 58.69 g/mol Boiling Pt: 2732 °C Melting Pt: 1453 °C Density: 8,90 Storage Temperature: Ambient |
MDL Number:
MFCD00011137 CAS Number: 7440-02-0 EINECS: 231-111-4 UN: 3089 ADR: 4.1,III Merck Index: 12,06582 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...