Chromium ≥99.996%, crystallites
Supplier: STREM CHEMICALS INC MS
24-0025-50G
24-0025-10G
100208-860EA
169.85
USD
100208-860
100204-538
Chromium ≥99.996%, crystallites
Chromium
Formula:
Cr MW: 52 g/mol Boiling Pt: 2200 °C Melting Pt: 1890 °C Density: 7.20 |
MDL Number:
MFCD00010944 CAS Number: 7440-47-3 EINECS: 231-157-5 UN: 3089 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...