tri-Calcium phosphate (∼37% Ca)
Supplier: STREM CHEMICALS INC MS
Synonyms:
Calcium phosphate tribasic
, penta-Calcium hydroxide triphosphate
, Tricalcium orthophosphate
, Calcium phosphate
20-2039-1000G
20-2039-250G
20-2039-5000G
100201-518EA
36.8
USD
100201-518
100200-888
100208-848
tri-Calcium phosphate (∼37% Ca)
tri-Calcium phosphate
Formula:
Ca₃(PO₄)₂ MW: 310.18 g/mol Density: 3.14 g/cm³ (20 °C) |
MDL Number:
MFCD00015984 CAS Number: 7758-87-4 EINECS: 231-840-8 Merck Index: 12,01741 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...