Calcium bromide, anhydrous ≥99.5% (Ca basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
20-1500-50G
20-1500-250G
100209-326EA
597.29
USD
100209-326
100199-364
Calcium bromide, anhydrous ≥99.5% (Ca basis)
Calcium bromide
Formula:
Br₂Ca MW: 199.89 g/mol Boiling Pt: 806…812 °C Melting Pt: ~ 730 °C Density: 3.353 |
MDL Number:
MFCD00010902 CAS Number: 7789-41-5 EINECS: 232-164-6 Merck Index: 12,01693 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...