Tripotassium orthophosphate, anhydrous ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Potassium phosphatePotassium phosphate tribasic

19-3800-250G 19-3800-1000G
100198-694EA 110.4 USD
100198-694 100209-760
Tripotassium orthophosphate, anhydrous ≥97%
tri-Potassium orthophosphate
Formula: K₃O₄P
MW: 212.27 g/mol
Melting Pt: 1340 °C
Density: 2.564 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036295
CAS Number: 7778-53-2
EINECS: 231-907-1
Merck Index: 12,07830

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR