Tris(3-methoxyphenyl)phosphine ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
15-7975-5G
15-7975-1G
100197-086EA
145.78
USD
100197-086
100197-088
Tris(3-methoxyphenyl)phosphine ≥97%
Tris(3-methoxyphenyl)phosphine
Phosphine
Formula:
C₂₁H₂₁O₃P MW: 352.37 g/mol Melting Pt: 115 °C |
MDL Number:
MFCD00008386 CAS Number: 29949-84-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...