Triisopropyl phosphite ≥94%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Tri-i-propylphosphite
15-7000-250G
15-7000-50G
100211-026EA
260.43
USD
100211-026
100207-596
Triisopropyl phosphite ≥94%
Triisopropyl phosphite
Formula:
C₉H₂₁O₃P MW: 208.24 g/mol Boiling Pt: 63…64 °C (11 mmHg) Density: 0,844 Flash Pt: 154 °F |
MDL Number:
MFCD00008874 CAS Number: 116-17-6 EINECS: 204-130-0 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...