Trioctylphosphine ≥97%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Tri-n-octylphosphine
, TOP
15-6655-25G
15-6655-100G
15-6655-500G
100203-012EA
547.75
USD
100203-012
100203-010
100198-674
Trioctylphosphine ≥97%
Trioctylphosphine
Nanomaterials, Phosphine, Surfactants for Nano Synthesis
Formula:
C₂₄H₅₁P MW: 370.64 g/mol Boiling Pt: 175 °C (0.3 mmHg) Melting Pt: 30 °C Density: 0,83 Flash Pt: 280 °F |
MDL Number:
MFCD00015298 CAS Number: 4731-53-7 EINECS: 225-234-2 UN: 1760 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...