Trimethyl phosphite ≥97%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
TMP
15-6550-1000G
15-6550-250G
100206-434EA
90.58
USD
100206-434
100206-432
Trimethyl phosphite ≥97%
Trimethyl phosphite
Formula:
C₃H₉O₃P Boiling Pt: 110…112 °C Melting Pt: –77.8 °C Density: 1,052 Flash Pt: 82 °F |
MDL Number:
MFCD00008350 CAS Number: 121-45-9 EINECS: 204-471-5 UN: 2329 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...