(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S)-(-)-BINAP(S)-(-)-2,2'-Bis(diphenylphoshino)-1,1'-binaphthyl(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene(S)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl

15-0151-250MG 15-0151-1G 15-0151-5G
100198-602EA 42.46 USD
100198-602 100211-000 100211-002
(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl ≥98%
(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl

BINAP, Phosphine

Formula: C₄₄H₃₂P₂
MW: 622.69 g/mol
Melting Pt: 241…242 °C
Storage Temperature: Ambient
MDL Number: MFCD00010805
CAS Number: 76189-56-5
Merck Index: 13,01223

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR